accessibility menu, dialog, popup

UserName

CAS RN 80194-68-9

IUPAC Name: 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
Synonyms: 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 225.55
Molecular Formula: C7H3ClF3NO2
InChi Key: HXRMCZBDTDCCOP-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=C(C=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results