missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-(trifluoromethyl)benzonitrile 98.0+%, TCI America™
$123.03 - $432.46
Chemical Identifiers
| CAS | 693245-52-2 |
|---|---|
| Molecular Formula | C8H3ClF3N |
| Molecular Weight (g/mol) | 205.56 |
| MDL Number | MFCD04115854 |
| InChI Key | RLHYYDKFOQSWIB-UHFFFAOYSA-N |
| Synonym | 3-chloro-5-trifluoromethyl benzonitrile, 5-chloro-3-cyanobenzotrifluoride, 3-chloro-5-trifluoromethyl-benzonitrile, 3-chloro-5-cyanobenzotrifluoride, benzonitrile, 3-chloro-5-trifluoromethyl, 5-chloro-3-cyano benzotrifluoride, pubchem16810, 5-chloro-3-trifluoromethyl benzenecarbonitrile |
| PubChem CID | 4584933 |
| IUPAC Name | 3-chloro-5-(trifluoromethyl)benzonitrile |
| SMILES | FC(F)(F)C1=CC(Cl)=CC(=C1)C#N |
Chemical Identifiers
| 693245-52-2 | |
| 205.56 | |
| RLHYYDKFOQSWIB-UHFFFAOYSA-N | |
| 4584933 | |
| FC(F)(F)C1=CC(Cl)=CC(=C1)C#N |
| C8H3ClF3N | |
| MFCD04115854 | |
| 3-chloro-5-trifluoromethyl benzonitrile, 5-chloro-3-cyanobenzotrifluoride, 3-chloro-5-trifluoromethyl-benzonitrile, 3-chloro-5-cyanobenzotrifluoride, benzonitrile, 3-chloro-5-trifluoromethyl, 5-chloro-3-cyano benzotrifluoride, pubchem16810, 5-chloro-3-trifluoromethyl benzenecarbonitrile | |
| 3-chloro-5-(trifluoromethyl)benzonitrile |
Specifications
| 693245-52-2 | |
| C8H3ClF3N | |
| 5 g | |
| 3-chloro-5-trifluoromethyl benzonitrile, 5-chloro-3-cyanobenzotrifluoride, 3-chloro-5-trifluoromethyl-benzonitrile, 3-chloro-5-cyanobenzotrifluoride, benzonitrile, 3-chloro-5-trifluoromethyl, 5-chloro-3-cyano benzotrifluoride, pubchem16810, 5-chloro-3-trifluoromethyl benzenecarbonitrile | |
| FC(F)(F)C1=CC(Cl)=CC(=C1)C#N | |
| 205.56 | |
| 205.56 | |
| Liquid |
| Colorless | |
| MFCD04115854 | |
| 3276 | |
| RLHYYDKFOQSWIB-UHFFFAOYSA-N | |
| 3-chloro-5-(trifluoromethyl)benzonitrile | |
| 4584933 | |
| ≥98.0% (GC) | |
| 3-Chloro-5-(trifluoromethyl)benzonitrile |
Safety and Handling
TSCA : No