missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-5-(trifluoromethyl)benzene-1,2-diamine, 97%, Thermo Scientific™

Catalog No. btb05127ea
Change view
Click to view available options
Quantity:
10g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB05127EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB05127EA Supplier Thermo Scientific Chemicals Supplier No. BTB05127EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132915-80-1
Molecular Formula C7H6ClF3N2
Molecular Weight (g/mol) 210.58
MDL Number MFCD00042198
InChI Key NVNLHLRVSHJKBN-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl benzene-1,2-diamine, 3-chloro-4,5-diaminobenzotrifluoride, 1,2-diamino-3-chloro-5-trifluoromethyl benzene, 3-chloro-5-trifluoromethyl-1,2-phenylenediamine, 3-chloro-5-trifluoromethyl-1,2-benzenediamine, timtec-bb sbb003446, buttpark 75\04-15, 1,2-diamino-3-chloro-5trifluoromethylbenzene
PubChem CID 735784
IUPAC Name 3-chloro-5-(trifluoromethyl)benzene-1,2-diamine
SMILES NC1=CC(=CC(Cl)=C1N)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10g
Formula Weight 210.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzene-1, 2-diamine
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.