Learn More
3-Chloro-5-methylaniline, 95%
CAS: 29027-20-1 | C7H8ClN | 141.60 g/mol
Supplier: Thermo Scientific Chemicals H6651806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Chloro-5-methylaniline | |
| C7H8ClN | |
| 5 g | |
| 742113 | |
| 3-chloro-5-methylbenzenamine, 3-amino-5-chlorotoluene, 3-chloro-5-methyl-phenylamine, benzenamine, 3-chloro-5-methyl, 3-chloro-5-methyl-aniline, 3-chloro-5-methylphenylamine, ksc497k2p | |
| CC1=CC(N)=CC(Cl)=C1 | |
| 141.60 | |
| 141.6 | |
| Solid |
| 29027-20-1 | |
| MFCD15527197 | |
| UN2239 | |
| Light sensitive | |
| XUZCJDBXXYJXDD-UHFFFAOYSA-N | |
| 3-chloro-5-methylaniline | |
| 12475434 | |
| 95% |
Chemical Identifiers
| 29027-20-1 | |
| 141.60 | |
| XUZCJDBXXYJXDD-UHFFFAOYSA-N | |
| 12475434 | |
| CC1=CC(N)=CC(Cl)=C1 |
| C7H8ClN | |
| MFCD15527197 | |
| 3-chloro-5-methylbenzenamine, 3-amino-5-chlorotoluene, 3-chloro-5-methyl-phenylamine, benzenamine, 3-chloro-5-methyl, 3-chloro-5-methyl-aniline, 3-chloro-5-methylphenylamine, ksc497k2p | |
| 3-chloro-5-methylaniline |
Safety and Handling
GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H373
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: CHLOROTOLUIDINES, SOLID
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only