missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-methylaniline, 95%

CAS: 29027-20-1 | C7H8ClN | 141.60 g/mol

Supplier:  Thermo Scientific Chemicals H6651803

 View more versions of this product

Catalog No. AAH6651803


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloro-5-methylaniline
C7H8ClN
1 g
742113
3-chloro-5-methylbenzenamine, 3-amino-5-chlorotoluene, 3-chloro-5-methyl-phenylamine, benzenamine, 3-chloro-5-methyl, 3-chloro-5-methyl-aniline, 3-chloro-5-methylphenylamine, ksc497k2p
CC1=CC(N)=CC(Cl)=C1
141.60
141.6
Solid
29027-20-1
MFCD15527197
UN2239
Light sensitive
XUZCJDBXXYJXDD-UHFFFAOYSA-N
3-chloro-5-methylaniline
12475434
95%

Chemical Identifiers

29027-20-1
141.60
XUZCJDBXXYJXDD-UHFFFAOYSA-N
12475434
CC1=CC(N)=CC(Cl)=C1
C7H8ClN
MFCD15527197
3-chloro-5-methylbenzenamine, 3-amino-5-chlorotoluene, 3-chloro-5-methyl-phenylamine, benzenamine, 3-chloro-5-methyl, 3-chloro-5-methyl-aniline, 3-chloro-5-methylphenylamine, ksc497k2p
3-chloro-5-methylaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H373

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: CHLOROTOLUIDINES, SOLID

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only