missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-methoxybenzoic acid, 98%

Catalog No. AAH3196006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3196006 5 g
AAH3196003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3196006 Supplier Thermo Scientific Chemicals Supplier No. H3196006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82477-67-6 | C8H7ClO3 | 186.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82477-67-6
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD01317994
InChI Key RRVHCVOVXPMKGF-UHFFFAOYSA-N
Synonym 3-chloro-5-methoxy-benzoic acid, 5-chloro-m-anisic acid, 3-carboxy-5-chloroanisole, 3-chloro-5-methoxybenzoicacid, benzoic acid, 3-chloro-5-methoxy, pubchem14017, acmc-20ac57, ksc447q4j, cs-waa0107, benzoicacid, 3-chloro-5-methoxy
PubChem CID 12909341
IUPAC Name 3-chloro-5-methoxybenzoic acid
SMILES COC1=CC(Cl)=CC(=C1)C(O)=O

Specifications

Quantity 5 g
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 3-Chloro-5-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only