missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluoropyridine 98.0+%, TCI America™

Catalog No. C30551G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30551G Supplier TCI America Supplier No. C30551G

Chemical Identifiers

CAS 514797-99-0
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.534
MDL Number MFCD05663707
InChI Key LTILYPOSFJLPSK-UHFFFAOYSA-N
Synonym 5-chloro-3-fluoropyridine, 3-fluoro-5-chloropyridine, 3-chloro-5-fluoro-pyridine, pyridine, 3-chloro-5-fluoro, pubchem6620, acmc-1axgq, pyridine,3-chloro-5-fluoro, 3-chloro-5-fluoropyridine, abbypharma ap-14-5727
PubChem CID 12031536
IUPAC Name 3-chloro-5-fluoropyridine
SMILES C1=C(C=NC=C1Cl)F

Specifications

Melting Point 26°C
Color White-Yellow
Boiling Point 134°C
Quantity 1 g
UN Number 1325
Formula Weight 131.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chloro-5-fluoropyridine
TSCA No
Recommended Storage Refrigerator