accessibility menu, dialog, popup

UserName

NClF

CAS RN 514797-99-0

IUPAC Name: 3-chloro-5-fluoropyridine
Synonyms: 3-chloro-5-fluoropyridine
Molecular Weight (g/mol): 131.53
Molecular Formula: C5H3ClFN
InChi Key: LTILYPOSFJLPSK-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CN=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)