missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-fluoropyridine, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 25°C to 27°C |
| Flash Point | 41°C (106°F) |
| Refractive Index | 1.5015 |
| Quantity | 1 g |
| UN Number | UN1325 |
| Formula Weight | 131.54 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Chloro-5-fluoropyridine |
Safety and Handling
| Hazard Category | H228-H302+H312+H332-H315-H318-H335 |
| Hazard Statement | GHS H Statement H318-H228-H302-H312-H332-H335-H315 Causes serious eye damage. Flammable solid. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only