missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorophenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific™

Catalog No. AAH54775AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54775AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54775AE Supplier Thermo Scientific Chemicals Supplier No. H54775AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Reacts with water.

Notes
Store at 4°C.

Chemical Identifiers

CAS 480438-50-4
Molecular Formula C6H3BrClFMg
Molecular Weight (g/mol) 233.75
MDL Number MFCD02260214
InChI Key IKMSSONTWSEQMT-UHFFFAOYSA-M
Synonym 3-chloro-5-fluorophenylmagnesium bromide, bromo 3-chloro-5-fluorophenyl magnesium, ikmssontwseqmt-uhfffaoysa-m, magnesium,bromo 3-chloro-5-fluorophenyl, magnesium, bromo 3-chloro-5-fluorophenyl, 3-chloro-5-fluorophenylmagnesium bromide solution, 3-chloro-5-fluorophenylmagnesium bromide, 0.50 m in 2-methf, 3-chloro-5-fluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3-chloro-5-fluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 2778633
IUPAC Name magnesium;1-chloro-3-fluorobenzene-5-ide;bromide
SMILES FC1=CC(Cl)=CC([Mg]Br)=C1

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 233.75
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 3-Chloro-5-fluorophenylmagnesium bromide
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon