missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$80.85 - $80.85
Specifications
| CAS | 328956-61-2 |
|---|---|
| Color | White |
| Molecular Formula | C6H5BClFO2 |
| MDL Number | MFCD06801741 |
| InChI Key | XYQDHVBKUNNLGZ-UHFFFAOYSA-N |
Specifications
| 328956-61-2 | |
| C6H5BClFO2 | |
| XYQDHVBKUNNLGZ-UHFFFAOYSA-N | |
| 15391287 | |
| Crystalline Powder |
| White | |
| MFCD06801741 | |
| (3-chloro-5-fluorophenyl)boronic acid | |
| 174.36 |