missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C29141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29141G Supplier TCI America Supplier No. C29141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Chloro-5-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 328956-61-2
Color White
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD06801741
Synonym 3-chloro-5-fluorobenzeneboronic acid, 3-chloro-5-fluorophenyl boronic acid, boronic acid, 3-chloro-5-fluorophenyl, pubchem6363, acmc-1agjy, ksc497k5n, 3-borono-5-fluorochlorobenzene, 3-chloro-5-fluoro-benzeneboronic acid, 3-chloro-5-fluoro-phenyl boronic acid, 3-chloranyl-5-fluoranyl-phenyl boronic acid
InChI Key XYQDHVBKUNNLGZ-UHFFFAOYSA-N
SMILES OB(O)C1=CC(F)=CC(Cl)=C1
IUPAC Name (3-chloro-5-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.36
PubChem CID 15391287
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less