missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorophenol, 98%

Catalog No. AAH2619503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2619503 1 g
AAH2619506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2619503 Supplier Thermo Scientific Chemicals Supplier No. H2619503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 202982-70-5 | C6H4ClFO | 146.545 g/mol

3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 202982-70-5
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
MDL Number MFCD00042212
InChI Key ZMFBYTAAMHWQHD-UHFFFAOYSA-N
Synonym 3-chloro-5-fluoro-phenol, phenol, 3-chloro-5-fluoro, pubchem2858, acmc-209f8b, 3-chloranyl-5-fluoranyl-phenol, phenol, 3-chloro-5-fluoro-9ci
PubChem CID 2773713
IUPAC Name 3-chloro-5-fluorophenol
SMILES C1=C(C=C(C=C1F)Cl)O

Specifications

Refractive Index 1.537
Quantity 1 g
UN Number UN3265
Solubility Information Immiscible with water.
Formula Weight 146.55
Percent Purity 98%
Chemical Name or Material 3-Chloro-5-fluorophenol
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only