missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. p-7054181
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62180MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH62180MD Supplier Thermo Scientific Chemicals Supplier No. H62180MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1245524-02-0
Molecular Formula C12H15BClFO2
Molecular Weight (g/mol) 256.508
InChI Key ZYWCNKHMEUXNGS-UHFFFAOYSA-N
Synonym 2-3-chloro-5-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-chloro-5-fluoro-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, 3-chloro-5-fluorophenylboronic acid pinacol ester, 3-chloro-5-fluorobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-chloro-5-fluorophenyl-4,4,5,5-tetramethyl
PubChem CID 53416044
IUPAC Name 2-(3-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)Cl)F

Specifications

Quantity 250 mg
Formula Weight 256.51
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Chloro-5-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures