missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzeneboronic acid, 95%

Catalog No. AAH3213703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3213703 1 g
AAH3213706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3213703 Supplier Thermo Scientific Chemicals Supplier No. H3213703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 328956-61-2 | C6H5BClFO2 | 174.36 g/mol

3-Chloro-5-fluorobenzeneboronic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-5-fluorobenzeneboronic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 328956-61-2
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD06801741
InChI Key XYQDHVBKUNNLGZ-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobenzeneboronic acid, 3-chloro-5-fluorophenyl boronic acid, boronic acid, 3-chloro-5-fluorophenyl, pubchem6363, acmc-1agjy, ksc497k5n, 3-borono-5-fluorochlorobenzene, 3-chloro-5-fluoro-benzeneboronic acid, 3-chloro-5-fluoro-phenyl boronic acid, 3-chloranyl-5-fluoranyl-phenyl boronic acid
PubChem CID 15391287
IUPAC Name (3-chloro-5-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=CC(Cl)=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 174.37
Percent Purity 95%
Chemical Name or Material 3-Chloro-5-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Ambient temperatures

RUO – Research Use Only