missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-5-fluoroanisole 96.0+%, TCI America™
$87.60 - $350.44
Chemical Identifiers
| CAS | 202925-08-4 |
|---|---|
| Molecular Formula | C7H6ClFO |
| Molecular Weight (g/mol) | 160.57 |
| MDL Number | MFCD00070770 |
| InChI Key | XPZBNEWAZPZUHF-UHFFFAOYSA-N |
| PubChem CID | 2779258 |
| IUPAC Name | 1-chloro-3-fluoro-5-methoxybenzene |
| SMILES | COC1=CC(F)=CC(Cl)=C1 |
Chemical Identifiers
| 202925-08-4 | |
| 160.57 | |
| XPZBNEWAZPZUHF-UHFFFAOYSA-N | |
| 1-chloro-3-fluoro-5-methoxybenzene |
| C7H6ClFO | |
| MFCD00070770 | |
| 2779258 | |
| COC1=CC(F)=CC(Cl)=C1 |
Specifications
| 202925-08-4 | |
| 189°C | |
| MFCD00070770 | |
| 3-chloro-5-fluoroanisole, benzene,1-chloro-3-fluoro-5-methoxy, 1-chloro-3-fluoro-5-methoxy-benzene, pubchem2661, 3-chloro-5-fluoroanisol, 3-fluoro-5-methoxy-1-chlorobenzene, benzene, 1-chloro-3-fluoro-5-methoxy, 1-chloranyl-3-fluoranyl-5-methoxy-benzene | |
| COC1=CC(F)=CC(Cl)=C1 | |
| 160.57 | |
| 160.57 | |
| Liquid |
| Yellow | |
| C7H6ClFO | |
| 1 g | |
| XPZBNEWAZPZUHF-UHFFFAOYSA-N | |
| 1-chloro-3-fluoro-5-methoxybenzene | |
| 2779258 | |
| ≥96.0% (GC) | |
| 3-Chloro-5-fluoroanisole |
Safety and Handling
TSCA : No