missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethyl)benzeneboronic acid, 97%

CAS: 847756-88-1 | C7H5BClF3O2 | 224.37 g/mol

$78.38 - $217.14

Chemical Identifiers

CAS 847756-88-1
Molecular Formula C7H5BClF3O2
Molecular Weight (g/mol) 224.37
MDL Number MFCD03788417
InChI Key OCHKTAGGJMWISO-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethyl phenylboronic acid, 3-chloro-4-trifluoromethylphenylboronic acid, 3-chloro-4-trifluoromethyl benzeneboronic acid, 3-chloro-4-trifluoromethyl phenyl boronic acid, 4-borono-2-chlorobenzotrifluoride, boronic acid, 3-chloro-4-trifluoromethyl phenyl, pubchem23707, acmc-209pwg
PubChem CID 3772958
IUPAC Name [3-chloro-4-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)C(F)(F)F)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH53120MD
View Documents
Thermo Scientific Chemicals
H53120MD
250 mg
Each for $78.38
Only null left
 
AAH5312003
View Documents
Thermo Scientific Chemicals
H5312003
1 g
Each for $217.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

847756-88-1
224.37
OCHKTAGGJMWISO-UHFFFAOYSA-N
3772958
B(C1=CC(=C(C=C1)C(F)(F)F)Cl)(O)O
C7H5BClF3O2
MFCD03788417
3-chloro-4-trifluoromethyl phenylboronic acid, 3-chloro-4-trifluoromethylphenylboronic acid, 3-chloro-4-trifluoromethyl benzeneboronic acid, 3-chloro-4-trifluoromethyl phenyl boronic acid, 4-borono-2-chlorobenzotrifluoride, boronic acid, 3-chloro-4-trifluoromethyl phenyl, pubchem23707, acmc-209pwg
[3-chloro-4-(trifluoromethyl)phenyl]boronic acid

Specifications

847756-88-1
C7H5BClF3O2
250 mg
OCHKTAGGJMWISO-UHFFFAOYSA-N
[3-chloro-4-(trifluoromethyl)phenyl]boronic acid
3772958
97%
206°C to 212°C
MFCD03788417
3-chloro-4-trifluoromethyl phenylboronic acid, 3-chloro-4-trifluoromethylphenylboronic acid, 3-chloro-4-trifluoromethyl benzeneboronic acid, 3-chloro-4-trifluoromethyl phenyl boronic acid, 4-borono-2-chlorobenzotrifluoride, boronic acid, 3-chloro-4-trifluoromethyl phenyl, pubchem23707, acmc-209pwg
B(C1=CC(=C(C=C1)C(F)(F)F)Cl)(O)O
224.37
224.38
3-Chloro-4-(trifluoromethyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only