missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenyl Isocyanate 97.0+%, TCI America™

Supplier:  TCI America I05635G

 View more versions of this product

Catalog No. I05635G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Chloro-4-methylphenyl Isocyanate
White-Yellow
5 g
C8H6ClNO
3428
UKTKKMZDESVUEE-UHFFFAOYSA-N
2-chloro-4-isocyanato-1-methylbenzene
62832
≥97.0% (GC)
28479-22-3
124°C
Crystal-Lump at 20°C
MFCD00013858
3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
CC1=CC=C(C=C1Cl)N=C=O
167.59
167.59

Chemical Identifiers

28479-22-3
167.59
UKTKKMZDESVUEE-UHFFFAOYSA-N
62832
CC1=CC=C(C=C1Cl)N=C=O
C8H6ClNO
MFCD00013858
3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
2-chloro-4-isocyanato-1-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions