missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenyl Isocyanate 97.0+%, TCI America™

Catalog No. p-7131486
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05635G 5 g
I056325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05635G Supplier TCI America Supplier No. I05635G
Only null left

Chemical Identifiers

CAS 28479-22-3
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00013858
InChI Key UKTKKMZDESVUEE-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
PubChem CID 62832
IUPAC Name 2-chloro-4-isocyanato-1-methylbenzene
SMILES CC1=CC=C(C=C1Cl)N=C=O

Specifications

Color White-Yellow
Quantity 5 g
Physical Form Crystal-Lump at 20°C
UN Number 3428
Boiling Point 124°C
Formula Weight 167.59
Percent Purity ≥97.0% (GC)
Chemical Name or Material 3-Chloro-4-methylphenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator