missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenyl isocyanate, 98%

CAS: 28479-22-3 | C8H6ClNO | 167.59 g/mol

Supplier:  Thermo Scientific Chemicals L1206406

 View more versions of this product

Catalog No. AAL1206406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloro-4-methylphenyl isocyanate
28479-22-3
225°C
1.556
MFCD00013858
2690771
UKTKKMZDESVUEE-UHFFFAOYSA-N
2-chloro-4-isocyanato-1-methylbenzene
62832
98%
20°C to 24°C
1.224
5 g
C8H6ClNO
UN2236
3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
CC1=CC=C(C=C1Cl)N=C=O
167.59
167.6

Chemical Identifiers

28479-22-3
167.59
UKTKKMZDESVUEE-UHFFFAOYSA-N
62832
CC1=CC=C(C=C1Cl)N=C=O
C8H6ClNO
MFCD00013858
3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
2-chloro-4-isocyanato-1-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H314-H318
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P284-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H330-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-CHLORO-4-METHYLPHENYL ISOCYANATE, LIQUID

EINECSNumber : 249-050-7

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only