missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylphenyl isocyanate, 98%

Catalog No. p-7045811
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1206403 1 g
AAL1206406 5 g
AAL1206414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1206403 Supplier Thermo Scientific Chemicals Supplier No. L1206403

CAS: 28479-22-3 | C8H6ClNO | 167.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28479-22-3
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00013858
InChI Key UKTKKMZDESVUEE-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
PubChem CID 62832
IUPAC Name 2-chloro-4-isocyanato-1-methylbenzene
SMILES CC1=CC=C(C=C1Cl)N=C=O

Specifications

Melting Point 20°C to 24°C
Density 1.224
Quantity 1 g
Refractive Index 1.556
UN Number UN2236
Beilstein 2690771
Boiling Point 225°C
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-methylphenyl isocyanate
Hazard Category H314-H330-H334-H335
Hazard Statement GHS H Statement
H330-H334-H314-H318
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P284-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-CHLORO-4-METHYLPHENYL ISOCYANATE, LIQUID
EINECSNumber 249-050-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only