missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylbenzeneboronic acid, 97%

CAS: 175883-63-3 | C7H8BClO2 | 170.40 g/mol

$42.89 - $143.34

Chemical Identifiers

CAS 175883-63-3
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.40
MDL Number MFCD04039010
InChI Key YTJUYWRCAZWVSX-UHFFFAOYSA-N
Synonym 3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
PubChem CID 3854610
IUPAC Name (3-chloro-4-methylphenyl)boronic acid
SMILES CC1=CC=C(C=C1Cl)B(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2828803
View Documents
Thermo Scientific Chemicals
H2828803
1 g
Each for $42.89
Only null left
 
AAH2828806
View Documents
Thermo Scientific Chemicals
H2828806
5 g
Each for $143.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

175883-63-3
170.40
YTJUYWRCAZWVSX-UHFFFAOYSA-N
3854610
CC1=CC=C(C=C1Cl)B(O)O
C7H8BClO2
MFCD04039010
3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
(3-chloro-4-methylphenyl)boronic acid

Specifications

224°C to 230°C
1 g
MFCD04039010
YTJUYWRCAZWVSX-UHFFFAOYSA-N
(3-chloro-4-methylphenyl)boronic acid
3854610
97%
175883-63-3
C7H8BClO2
3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
CC1=CC=C(C=C1Cl)B(O)O
170.40
170.4
3-Chloro-4-methylbenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only