missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methoxyphenylacetic Acid 97.0+%, TCI America™

Catalog No. C33111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33111G 1 g
C33115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33111G Supplier TCI America Supplier No. C33111G

Chemical Identifiers

CAS 13721-20-5
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.62
MDL Number MFCD02664697
InChI Key OARWXDXELDVNOC-UHFFFAOYSA-N
PubChem CID 4681662
IUPAC Name 2-(3-chloro-4-methoxyphenyl)acetic acid
SMILES COC1=C(Cl)C=C(CC(O)=O)C=C1

Specifications

Melting Point 95°C
Color White
Quantity 1 g
Formula Weight 200.62
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-methoxyphenylacetic Acid