missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-4'-methoxyacetophenone 98.0+%, TCI America™

Catalog No. p-7125825
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30931G 1 g
C30935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30931G Supplier TCI America Supplier No. C30931G
Only null left

Chemical Identifiers

CAS 37612-52-5
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD04973413
InChI Key QILWOKAXHOAFOF-UHFFFAOYSA-N
Synonym 3-chloro-4-methoxyacetophenone, 1-3-chloro-4-methoxyphenyl ethanone, 3'-chloro-4'-methoxyacetophenone, 1-3-chloro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-chloro-4-methoxyphenyl, pubchem16367, 3-chloro-4-methoxy acetophenone, 3'-chloro-4'-methoxy acetophenone, 3-cl-4-ch3o-c6h3-coch3
PubChem CID 520857
IUPAC Name 1-(3-chloro-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)Cl

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 148°C
Quantity 1 g
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Chloro-4′-methoxyacetophenone