missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. c29691g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29691G Supplier TCI America Supplier No. C29691G

Specifications

Chemical Name or Material 3-Chloro-4-hydroxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 182344-13-4
Molecular Formula C6H6BClO3
Quantity 1 g
MDL Number MFCD09258739
Synonym 3-chloro-4-hydroxyphenyl boronic acid, 3-chloro-4-hydroxybenzeneboronic acid, 4-borono-2-chlorophenol, boronic acid, 3-chloro-4-hydroxyphenyl, acmc-1bwc3, 3-chloro-4-hydroxyphenylboronicacid, 3-chloro-4-hydroxy-phenylboronic acid, 3-chloro-4-hydroxy-phenyl boronic acid, 3-chloranyl-4-oxidanyl-phenyl boronic acid
InChI Key WWQIKFZZILXJHG-UHFFFAOYSA-N
SMILES OB(O)C1=CC(Cl)=C(O)C=C1
IUPAC Name (3-chloro-4-hydroxyphenyl)boronic acid
Molecular Weight (g/mol) 172.37
PubChem CID 22125373
Formula Weight 172.37
Physical Form Crystalline Powder
Show More Show Less