missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxyacetanilide, 98%

Catalog No. p-4812929
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450080010 1 g
AC450080050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450080010 Supplier Thermo Scientific Chemicals Supplier No. 450080010

CAS: 3964-54-3 | C8H8ClNO2 | 185.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3964-54-3
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD01318123
InChI Key JULKJDRBSRRBHT-UHFFFAOYSA-N
Synonym n-3-chloro-4-hydroxyphenyl acetamide, 3-chloro-4-hydroxyacetanilide, 4-acetylamino-2-chlorophenol, 2-chloro-4-acetamidophenol, acetamide, n-3-chloro-4-hydroxyphenyl, 3'-chloro-4'-hydroxyacetanilide, zlchem 1169, paracetamol impurity c, chloro-4-acetamidophenol, 3-chloro-4-hydroxyacet anilide
PubChem CID 77579
IUPAC Name N-(3-chloro-4-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=C(O)C(Cl)=C1

Specifications

Melting Point 144.0°C to 146.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in hot water
Formula Weight 185.61
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-hydroxyacetanilide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 223-570-4

RUO – Research Use Only