missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorothiophenol, 97%

Catalog No. AAA1085603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1085603 1 g
AAA1085606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1085603 Supplier Thermo Scientific Chemicals Supplier No. A1085603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 60811-23-6 | C6H3ClFS | 161.60 g/mol

Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharamaceutical intermediate.

Solubility
Sparingly Soluble (0.014 g/L) (25°C).

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 60811-23-6
Molecular Formula C6H3ClFS
Molecular Weight (g/mol) 161.60
MDL Number MFCD00052938
InChI Key SFSHSIFYTWIGSF-UHFFFAOYSA-M
Synonym 3-chloro-4-fluorothiophenol, 3-chloro-4-fluorobenzene-1-thiol, 3-chloro-4-fluoro-benzenethiol, pubchem2857, acmc-209wz2, 3-chloro-4-flurothiophenol, benzenethiol, 3-chloro-4-fluoro
PubChem CID 2734197
IUPAC Name 3-chloro-4-fluorobenzenethiol
SMILES FC1=CC=C([S-])C=C1Cl

Specifications

Boiling Point 68°C to 70°C (2 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble (0.014g/L) (25°C).
Formula Weight 162.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-fluorothiophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only