missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%

Catalog No. AAA1017506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1017506 5 g
AAA1017514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1017506 Supplier Thermo Scientific Chemicals Supplier No. A1017506
Only null left

CAS: 175135-74-7 | C6H7Cl2FN2 | 197.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175135-74-7
Molecular Formula C6H7Cl2FN2
Molecular Weight (g/mol) 197.034
MDL Number MFCD00052267
InChI Key DWCDDXINAZHRLK-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorophenylhydrazine hydrochloride, 3-chloro-4-fluorophenyl hydrazine hydrochloride, 1-3-chloro-4-fluorophenyl hydrazine hydrochloride, hydrazine, 3-chloro-4-fluorophenyl-, monohydrochloride, pubchem3507, acmc-1c5q6, labotest-bb lt01147590, ksc494o5j, 3-chloro-4-fluorophenylhydrazinehcl, 3-chloro-4-fluorophenylhydrazine hcl
PubChem CID 44181777
IUPAC Name (3-chloro-4-fluorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1NN)Cl)F.Cl

Specifications

Melting Point ∼210°C (decomposition)
Quantity 5 g
Formula Weight 197.04
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorophenylhydrazine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only