missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$55.84 - $55.84
Specifications
| CAS | 144432-85-9 |
|---|---|
| Melting Point | 247°C |
| Color | White-Yellow |
| Molecular Formula | C6H5BClFO2 |
| MDL Number | MFCD00051800 |
Specifications
| 144432-85-9 | |
| White-Yellow | |
| MFCD00051800 | |
| B(C1=CC(=C(C=C1)F)Cl)(O)O | |
| 174.362 | |
| 174.36 |
| 247°C | |
| C6H5BClFO2 | |
| WJDZZXIDQYKVDG-UHFFFAOYSA-N | |
| (3-chloro-4-fluorophenyl)boronic acid | |
| 2734660 | |
| Crystalline Powder |