missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C17601G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17601G Supplier TCI America Supplier No. C17601G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Chloro-4-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 144432-85-9
Melting Point 247°C
Color White-Yellow
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD00051800
Synonym 3-chloro-4-fluorobenzeneboronic acid, 3-chloro-4-fluorophenyl boronic acid, 3-chloro-4-fluorophenyl boranediol, 4-fluoro-3-chlorophenylboronic acid, 3-chloro-4-fluoro phenyl boronic acid, 3-chloro-4-fluoro-phenyl boronic acid, boronic acid, 3-chloro-4-fluorophenyl, b-3-chloro-4-fluorophenyl-boronic acid, pubchem1780
InChI Key WJDZZXIDQYKVDG-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)F)Cl)(O)O
IUPAC Name (3-chloro-4-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.362
PubChem CID 2734660
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less