missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluoronitrobenzene 95.0+%, TCI America™

Catalog No. C104125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C104125G 25 g
C1041250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C104125G Supplier TCI America Supplier No. C104125G

Chemical Identifiers

CAS 350-30-1
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.54
MDL Number MFCD00007206
InChI Key DPHCXXYPSYMICK-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoronitrobenzene, benzene, 2-chloro-1-fluoro-4-nitro, 3-chloro-4-fluoro-1-nitrobenzene, 4-fluoro-3-chloronitrobenzene, 1-chloro-2-fluoro-5-nitrobenzene, 2-chloro-1-fluoro-4-nitro-benzene, 3-chloro-4-fluoro-nitrobenzene, pubchem8539, acmc-209icj, dsstox_cid_29134
PubChem CID 67688
IUPAC Name 2-chloro-1-fluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(F)C(Cl)=C1

Specifications

Melting Point 43°C
Boiling Point 105°C
Quantity 25 g
UN Number 3077
Formula Weight 175.54
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-fluoronitrobenzene
EINECSNumber (3)-4071
RTECSNumber CZ0720000
TSCA No
Recommended Storage Refrigerator