missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-fluorobenzyl Bromide 98.0+%, TCI America™
$63.38 - $253.51
Chemical Identifiers
| CAS | 192702-01-5 |
|---|---|
| Molecular Formula | C7H5BrClF |
| Molecular Weight (g/mol) | 223.47 |
| MDL Number | MFCD01631551 |
| InChI Key | GGTQWWTYUKXFPP-UHFFFAOYSA-N |
| Synonym | 3-chloro-4-fluorobenzyl bromide, 4-bromomethyl-2-chloro-1-fluorobenzene, alpha-bromo-3-chloro-4-fluorotoluene, 1-bromomethyl-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobenzylbromide, 3-chloro-4-fluoro benzyl bromide, benzene, 4-bromomethyl-2-chloro-1-fluoro, pubchem2130, acmc-209ewf, 4-bromomethyl-2-chloro-1-fluoro-benzene |
| PubChem CID | 2757542 |
| IUPAC Name | 4-(bromomethyl)-2-chloro-1-fluorobenzene |
| SMILES | FC1=CC=C(CBr)C=C1Cl |
Chemical Identifiers
| 192702-01-5 | |
| 223.47 | |
| GGTQWWTYUKXFPP-UHFFFAOYSA-N | |
| 2757542 | |
| FC1=CC=C(CBr)C=C1Cl |
| C7H5BrClF | |
| MFCD01631551 | |
| 3-chloro-4-fluorobenzyl bromide, 4-bromomethyl-2-chloro-1-fluorobenzene, alpha-bromo-3-chloro-4-fluorotoluene, 1-bromomethyl-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobenzylbromide, 3-chloro-4-fluoro benzyl bromide, benzene, 4-bromomethyl-2-chloro-1-fluoro, pubchem2130, acmc-209ewf, 4-bromomethyl-2-chloro-1-fluoro-benzene | |
| 4-(bromomethyl)-2-chloro-1-fluorobenzene |
Specifications
| 192702-01-5 | |
| 65°C | |
| MFCD01631551 | |
| 3265 | |
| GGTQWWTYUKXFPP-UHFFFAOYSA-N | |
| 4-(bromomethyl)-2-chloro-1-fluorobenzene | |
| 2757542 | |
| ≥98.0% (GC) | |
| 3-Chloro-4-fluorobenzyl Bromide |
| Yellow | |
| C7H5BrClF | |
| 5 g | |
| 3-chloro-4-fluorobenzyl bromide, 4-bromomethyl-2-chloro-1-fluorobenzene, alpha-bromo-3-chloro-4-fluorotoluene, 1-bromomethyl-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobenzylbromide, 3-chloro-4-fluoro benzyl bromide, benzene, 4-bromomethyl-2-chloro-1-fluoro, pubchem2130, acmc-209ewf, 4-bromomethyl-2-chloro-1-fluoro-benzene | |
| FC1=CC=C(CBr)C=C1Cl | |
| 223.47 | |
| 223.47 | |
| Liquid |
Safety and Handling
TSCA : No