missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7125274
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20815G 5 g
C208125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20815G Supplier TCI America Supplier No. C20815G

Chemical Identifiers

CAS 117482-84-5
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.556
MDL Number MFCD00015431
InChI Key VAHXXQJJZKBZDX-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoro-benzonitrile, benzonitrile, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenecarbonitrile, pubchem1561, acmc-1bnmf, 3-chloro-4-florobenzonitrile, 3-chloro-4-fluorobenzontrile, ksc174m4n, 3-chloro,4-fluorobenzonitrile, 3-chloro4-fluorobenzo-nitrile
PubChem CID 145525
IUPAC Name 3-chloro-4-fluorobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Cl)F

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Boiling Point 135°C
Formula Weight 155.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Chloro-4-fluorobenzonitrile