accessibility menu, dialog, popup

UserName

FClN

CAS RN 117482-84-5

IUPAC Name: 3-chloro-4-fluorobenzonitrile
Synonyms: 3-chloro-4-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: VAHXXQJJZKBZDX-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1Cl)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)