missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzonitrile, 98+%

Catalog No. AAA1598714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1598714 25 g
AAA1598706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1598714 Supplier Thermo Scientific Chemicals Supplier No. A1598714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 117482-84-5 | C7H3ClFN | 155.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117482-84-5
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.556
MDL Number MFCD00015431
InChI Key VAHXXQJJZKBZDX-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoro-benzonitrile, benzonitrile, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenecarbonitrile, pubchem1561, acmc-1bnmf, 3-chloro-4-florobenzonitrile, 3-chloro-4-fluorobenzontrile, ksc174m4n, 3-chloro,4-fluorobenzonitrile, 3-chloro4-fluorobenzo-nitrile
PubChem CID 145525
IUPAC Name 3-chloro-4-fluorobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Cl)F

Specifications

Melting Point 68°C to 72°C
Quantity 25 g
UN Number UN3439
Beilstein 4740501
Formula Weight 155.56
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-4-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only