missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-cyanopyridine 98.0+%, TCI America™

Catalog No. C29981G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29981G Supplier TCI America Supplier No. C29981G
Only null left

Chemical Identifiers

CAS 68325-15-5
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.554
MDL Number MFCD05663706
InChI Key JLLJPPBGJVCFGG-UHFFFAOYSA-N
Synonym 3-chloro-4-cyanopyridine, 3-chloroisonicotinonitrile, 4-pyridinecarbonitrile, 3-chloro, 3-chloro-4-pyridinecarbonitrile, pubchem15941, 3-chloro-isonicotinonitrile, ksc352o9n, 3-chloro-4-pyridine carbonitrile, 4-pyridinecarbonitrile, 3-chloro-9ci, c.i. 45161:2; 3,6-bis ethylamino-9-2-methoxycarbonyl phenyl-2,7-dimethylxanthylium molybdatephosphate
PubChem CID 10678306
IUPAC Name 3-chloropyridine-4-carbonitrile
SMILES C1=CN=CC(=C1C#N)Cl

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 138.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-cyanopyridine