accessibility menu, dialog, popup

UserName

CAS RN 68325-15-5

NNCl

CAS RN 68325-15-5

IUPAC Name: 3-chloropyridine-4-carbonitrile
Synonyms: 3-chloropyridine-4-carbonitrile
Molecular Weight (g/mol): 138.55
Molecular Formula: C6H3ClN2
InChi Key: JLLJPPBGJVCFGG-UHFFFAOYSA-N
SMILES: ClC1=C(C=CN=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results