missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-cyanobenzeneboronic acid, 96%
CAS: 1008415-02-8 | C7H5BClNO2 | 181.382 g/mol
$358.36 - $1011.52
Chemical Identifiers
| CAS | 1008415-02-8 |
|---|---|
| Molecular Formula | C7H5BClNO2 |
| Molecular Weight (g/mol) | 181.382 |
| InChI Key | QCJTUOIMDNJEHR-UHFFFAOYSA-N |
| Synonym | 3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid |
| PubChem CID | 46738932 |
| IUPAC Name | (3-chloro-4-cyanophenyl)boronic acid |
| SMILES | B(C1=CC(=C(C=C1)C#N)Cl)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH61543MD
|
Thermo Scientific Chemicals
H61543MD |
250 mg |
Each for $358.36
|
|
|||||
|
AAH6154303
|
Thermo Scientific Chemicals
H6154303 |
1 g |
Each for $1,011.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1008415-02-8 | |
| 181.382 | |
| 3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid | |
| (3-chloro-4-cyanophenyl)boronic acid |
| C7H5BClNO2 | |
| QCJTUOIMDNJEHR-UHFFFAOYSA-N | |
| 46738932 | |
| B(C1=CC(=C(C=C1)C#N)Cl)(O)O |
Specifications
| 1008415-02-8 | |
| 250 mg | |
| QCJTUOIMDNJEHR-UHFFFAOYSA-N | |
| (3-chloro-4-cyanophenyl)boronic acid | |
| 46738932 | |
| 96% | |
| 3-Chloro-4-cyanobenzeneboronic acid |
| C7H5BClNO2 | |
| 3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid | |
| B(C1=CC(=C(C=C1)C#N)Cl)(O)O | |
| 181.382 | |
| 164.93 | |
| Solid |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only