missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-cyanobenzeneboronic acid, 96%

CAS: 1008415-02-8 | C7H5BClNO2 | 181.382 g/mol

Supplier:  Thermo Scientific Chemicals H6154303

 View more versions of this product

Catalog No. AAH6154303


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloro-4-cyanobenzeneboronic acid
C7H5BClNO2
3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid
B(C1=CC(=C(C=C1)C#N)Cl)(O)O
181.382
164.93
Solid
1008415-02-8
1 g
QCJTUOIMDNJEHR-UHFFFAOYSA-N
(3-chloro-4-cyanophenyl)boronic acid
46738932
96%

Chemical Identifiers

1008415-02-8
181.382
3-chloro-4-cyanophenyl boronic acid, boronic acid, b-3-chloro-4-cyanophenyl, acmc-2097sr, 3-chloro-4-cyanophenyl boronicacid
(3-chloro-4-cyanophenyl)boronic acid
C7H5BClNO2
QCJTUOIMDNJEHR-UHFFFAOYSA-N
46738932
B(C1=CC(=C(C=C1)C#N)Cl)(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only