missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(3-fluorobenzyloxy)nitrobenzene 98.0+%, TCI America™

Catalog No. C28755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28755G 5 g
C28751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28755G Supplier TCI America Supplier No. C28755G
Only null left

Chemical Identifiers

CAS 443882-99-3
Molecular Formula C13H9ClFNO3
Molecular Weight (g/mol) 281.667
MDL Number MFCD06809827
InChI Key SFTHVDYRPHJAND-UHFFFAOYSA-N
Synonym 2-Chloro-1-(3-fluorobenzyloxy)-4-nitrobenzene
PubChem CID 7059269
IUPAC Name 2-chloro-1-[(3-fluorophenyl)methoxy]-4-nitrobenzene
SMILES C1=CC(=CC(=C1)F)COC2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 281.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-(3-fluorobenzyloxy)nitrobenzene