missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-3-methyl-1-butyne, 95%

Catalog No. aab2525214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2525214 25 g
AAB2525206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2525214 Supplier Thermo Scientific Chemicals Supplier No. B2525214
Only null left

CAS: 1111-97-3 | C5H7Cl | 102.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1111-97-3
Molecular Formula C5H7Cl
Molecular Weight (g/mol) 102.56
MDL Number MFCD00190221
InChI Key QSILYWCNPOLKPN-UHFFFAOYSA-N
Synonym 3-chloro-3-methyl-1-butyne, 1-butyne, 3-chloro-3-methyl, 3-chloro-3-methylbutyne, 3-methyl-3-chlorobutyne, acmc-1bo5c, 2-chloro-2-methyl butyne, 3-chloro-3-methyl butyne, 3-chloro-3-methyl-butyne, 3-chloro-3-methyl-1-butyn, 2-chloro-2-methyl-3-butyne
PubChem CID 70679
IUPAC Name 3-chloro-3-methylbut-1-yne
SMILES CC(C)(Cl)C#C

Specifications

Melting Point -61°C
Density 0.913
Boiling Point 73°C to 75°C
Flash Point 4°C (40°F)
Odor Recognizable
Refractive Index 1.4178
Quantity 25 g
UN Number UN1993
Formula Weight 102.57
Percent Purity 95%
Chemical Name or Material 3-Chloro-3-methyl-1-butyne
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 214-188-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only