accessibility menu, dialog, popup

UserName

CH3CH3ClHC

CAS RN 1111-97-3

IUPAC Name: 3-chloro-3-methylbut-1-yne
Synonyms: 1-butyne, 3-chloro-3-methyl-
Molecular Weight (g/mol): 102.56
Molecular Formula: C5H7Cl
InChi Key: QSILYWCNPOLKPN-UHFFFAOYSA-N
SMILES: CC(C)(Cl)C#C
Boiling Point 
  • (4)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)