missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7124726
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09885G 5 g
C098825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09885G Supplier TCI America Supplier No. C09885G

Chemical Identifiers

CAS 4771-47-5
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
InChI Key VCHSXYHBMFKRBK-UHFFFAOYSA-N
Synonym 3-chloro-2-nitrobenzoic acid, benzoic acid, 3-chloro-2-nitro, 3-chloro-2-nitrobenzoicacid, 3-chloro-2-nitro-benzoic acid, chlornitrobenzoesaure, pubchem11043, chloro-2-nitrobenzoic acid, acmc-209ka6, 3-chloro2-nitrobenzoic acid, ksc237s1n
PubChem CID 20902
SMILES C1=CC(=C(C(=C1)Cl)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 239°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-nitrobenzoic Acid
EINECSNumber (3)-1502
TSCA No