missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylbenzeneboronic acid, 97%

CAS: 313545-20-9 | C7H8BClO2 | 170.399 g/mol

Supplier:  Thermo Scientific Chemicals H3089303

 View more versions of this product

Catalog No. AAH3089303


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloro-2-methylbenzeneboronic acid
1 g
MFCD04115641
BJPNVVXTUYMJPN-UHFFFAOYSA-N
(3-chloro-2-methylphenyl)boronic acid
3744103
97%
313545-20-9
C7H8BClO2
3-chloro-2-methylphenyl boronic acid, 2-borono-6-chlorotoluene, 3-chloro-2-methylbenzeneboronic acid, 3-chloro-2-methylphenyboronic acid, boronic acid, 3-chloro-2-methylphenyl, boronic acid,b-3-chloro-2-methylphenyl, 3-chloro-2-methyl-phenyl boronic acid, acmc-209hls, 3-chloro-2-methylphenboronic acid, 3-chloro-2-methyl-phenylboronic acid
B(C1=C(C(=CC=C1)Cl)C)(O)O
170.399
170.4

Chemical Identifiers

313545-20-9
170.399
BJPNVVXTUYMJPN-UHFFFAOYSA-N
3744103
B(C1=C(C(=CC=C1)Cl)C)(O)O
C7H8BClO2
MFCD04115641
3-chloro-2-methylphenyl boronic acid, 2-borono-6-chlorotoluene, 3-chloro-2-methylbenzeneboronic acid, 3-chloro-2-methylphenyboronic acid, boronic acid, 3-chloro-2-methylphenyl, boronic acid,b-3-chloro-2-methylphenyl, 3-chloro-2-methyl-phenyl boronic acid, acmc-209hls, 3-chloro-2-methylphenboronic acid, 3-chloro-2-methyl-phenylboronic acid
(3-chloro-2-methylphenyl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only