missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylbenzeneboronic acid, 97%

Catalog No. p-7030611
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH30893MD 250 mg
AAH3089303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH30893MD Supplier Thermo Scientific Chemicals Supplier No. H30893MD

CAS: 313545-20-9 | C7H8BClO2 | 170.399 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 313545-20-9
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.399
MDL Number MFCD04115641
InChI Key BJPNVVXTUYMJPN-UHFFFAOYSA-N
Synonym 3-chloro-2-methylphenyl boronic acid, 2-borono-6-chlorotoluene, 3-chloro-2-methylbenzeneboronic acid, 3-chloro-2-methylphenyboronic acid, boronic acid, 3-chloro-2-methylphenyl, boronic acid,b-3-chloro-2-methylphenyl, 3-chloro-2-methyl-phenyl boronic acid, acmc-209hls, 3-chloro-2-methylphenboronic acid, 3-chloro-2-methyl-phenylboronic acid
PubChem CID 3744103
IUPAC Name (3-chloro-2-methylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)Cl)C)(O)O

Specifications

Quantity 250 mg
Formula Weight 170.4
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-methylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only