missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylaniline 99.0+%, TCI America™

Catalog No. C030025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C030025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C030025ML Supplier TCI America Supplier No. C030025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 87-60-5
Melting Point 3°C
Color Brown
Boiling Point 245°C
Molecular Formula C7H8ClN
Quantity 25 mL
MDL Number MFCD00007766
UN Number 3429
Synonym 3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline
InChI Key ZUVPLKVDZNDZCM-UHFFFAOYSA-N
SMILES CC1=C(N)C=CC=C1Cl
IUPAC Name 3-chloro-2-methylaniline
Molecular Weight (g/mol) 141.60
PubChem CID 6894
Formula Weight 141.60
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-methylaniline
Show More Show Less
EINECSNumber (3)-0265
RTECSNumber XU4760000
TSCA Yes