Learn More
3-Chloro-2-methylaniline, 98+%
CAS: 87-60-5 | C7H8ClN | 141.60 g/mol
$213.60 - $797.92
Chemical Identifiers
| CAS | 87-60-5 |
|---|---|
| Molecular Formula | C7H8ClN |
| Molecular Weight (g/mol) | 141.60 |
| MDL Number | MFCD00007766 |
| InChI Key | ZUVPLKVDZNDZCM-UHFFFAOYSA-N |
| Synonym | 3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline |
| PubChem CID | 6894 |
| IUPAC Name | 3-chloro-2-methylaniline |
| SMILES | CC1=C(N)C=CC=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1822130
|
Thermo Scientific Chemicals
A1822130 |
250 g |
Each for $213.60
|
|
|||||
|
AAA182210B
|
Thermo Scientific Chemicals
A182210B |
1000 g |
Each for $797.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 87-60-5 | |
| 141.60 | |
| ZUVPLKVDZNDZCM-UHFFFAOYSA-N | |
| 6894 | |
| CC1=C(N)C=CC=C1Cl |
| C7H8ClN | |
| MFCD00007766 | |
| 3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline | |
| 3-chloro-2-methylaniline |
Specifications
| 87-60-5 | |
| 1.188 | |
| 113°C (235°F) | |
| 1.589 | |
| 250 g | |
| 471697 | |
| ZUVPLKVDZNDZCM-UHFFFAOYSA-N | |
| 3-chloro-2-methylaniline | |
| 6894 | |
| ≥98% |
| 2°C | |
| 245°C | |
| C7H8ClN | |
| MFCD00007766 | |
| UN3429 | |
| 3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline | |
| CC1=C(N)C=CC=C1Cl | |
| 141.60 | |
| 141.6 | |
| 3-Chloro-2-methylaniline |
Safety and Handling
GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P361-P363-P501c
H302-H311-H315-H318-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, LIQUID
EINECSNumber : 201-756-6
RTECSNumber : XU4760000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only