missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methyl-1-propene 98.0+%, TCI America™

Catalog No. p-7124282
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C020925ML 25 mL
C0209500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C020925ML Supplier TCI America Supplier No. C020925ML
Only null left

Chemical Identifiers

CAS 563-47-3
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00000953
InChI Key OHXAOPZTJOUYKM-UHFFFAOYSA-N
Synonym 3-chloro-2-methylpropene, methallyl chloride, 3-chloro-2-methyl-1-propene, 1-propene, 3-chloro-2-methyl, 2-methylallyl chloride, isobutenyl chloride, 2-methyl-2-propenyl chloride, beta-methallyl chloride, propene, 3-chloro-2-methyl, cloruro di metallile
PubChem CID 11241
ChEBI CHEBI:82419
IUPAC Name 3-chloro-2-methylprop-1-ene
SMILES CC(=C)CCl

Specifications

Color Colorless
Boiling Point 73°C
Quantity 25 mL
UN Number 2554
Formula Weight 90.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-methyl-1-propene
EINECSNumber (2)-0117&(2)-2367
RTECSNumber UC8050000
TSCA Yes