accessibility menu, dialog, popup

UserName

CAS RN 563-47-3

IUPAC Name: 3-chloro-2-methylprop-1-ene
Synonyms: 3-chloro-2-methylpropene
Molecular Weight (g/mol): 90.55
Molecular Formula: C4H7Cl
InChi Key: OHXAOPZTJOUYKM-UHFFFAOYSA-N
SMILES: CC(=C)CCl
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)