missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-hydroxypyridine 98.0+%, TCI America™

Catalog No. p-7125419
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C23401G 1 g
C23405G 5 g
2 options

Catalog No. C23401G

Supplier: TCI America C23401G

Only null left

Chemical Identifiers

CAS 13466-35-8
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD01646109
InChI Key XTYNIPUFKBBALX-UHFFFAOYSA-N
Synonym 3-chloro-2-hydroxypyridine, 3-chloropyridin-2-ol, 3-chloro-2-pyridinol, 2-hydroxy-3-chloropyridine, 3-chloropyridin-2 1h-one, 3-chloro-2 1h-pyridinone, chloropyridone, 2 1h-pyridinone, 3-chloro, chloropyridinol, pubchem6538
PubChem CID 582210
IUPAC Name 3-chloro-1,2-dihydropyridin-2-one
SMILES ClC1=CC=CNC1=O

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 1 g
Formula Weight 129.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-hydroxypyridine